The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105244.1 | NP_001098714.1 | 1 - 2 | 578 | 73 |
NM_002845.3 | NP_002836.3 | 1 - 2 | 578 | 73 | |
XM_011525708.2 | XP_011524010.1 | 1 - 2 | 573 | 73 | |
XM_011525710.2 | XP_011524012.1 | 1 - 2 | 573 | 73 | |
XM_011525712.2 | XP_011524014.1 | 1 - 2 | 573 | 73 | |
XM_017025911.1 | XP_016881400.1 | 1 - 2 | 573 | 73 | |
GenBank mRNA | AK303652.1 | - | 1 - 2 | 76 | 73 |
BC151842.1 | - | 1 - 2 | 1124 | 73 | |
DC381904.1 | - | 1 - 2 | 509 | 73 | |
X58288.1 | - | 1 - 2 | 75 | 73 |