Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRM
Entrez Gene ID
5797
Gene Name
protein tyrosine phosphatase, receptor type M
Gene Aliases
PTPRL1, R-PTP-MU, RPTPM, RPTPU, hR-PTPu
Chromosome Location
Chr.18: 7567316 - 8406861 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105244.1 NP_001098714.1 8 - 9 1941 67
NM_002845.3 NP_002836.3 8 - 9 1941 67
XM_011525708.2 XP_011524010.1 8 - 9 1936 67
XM_011525710.2 XP_011524012.1 8 - 9 1936 67
XM_011525712.2 XP_011524014.1 8 - 9 1936 67
XM_011525713.2 XP_011524015.1 8 - 9 1536 67
XM_011525715.2 XP_011524017.1 4 - 5 901 67
XM_011525716.2 XP_011524018.1 4 - 5 895 67
XM_011525717.2 XP_011524019.1 2 - 3 610 67
XM_011525722.2 XP_011524024.1 4 - 5 901 67
XM_017025894.1 XP_016881383.1 8 - 9 1480 67
XM_017025895.1 XP_016881384.1 8 - 9 1480 67
XM_017025896.1 XP_016881385.1 8 - 9 1480 67
XM_017025897.1 XP_016881386.1 8 - 9 1480 67
XM_017025898.1 XP_016881387.1 8 - 9 1480 67
XM_017025899.1 XP_016881388.1 8 - 9 1480 67
XM_017025900.1 XP_016881389.1 8 - 9 1480 67
XM_017025901.1 XP_016881390.1 8 - 9 1480 67
XM_017025902.1 XP_016881391.1 8 - 9 1480 67
XM_017025903.1 XP_016881392.1 8 - 9 1480 67
XM_017025904.1 XP_016881393.1 8 - 9 1480 67
XM_017025905.1 XP_016881394.1 8 - 9 1536 67
XM_017025906.1 XP_016881395.1 4 - 5 885 67
XM_017025907.1 XP_016881396.1 4 - 5 901 67
XM_017025908.1 XP_016881397.1 4 - 5 901 67
XM_017025909.1 XP_016881398.1 4 - 5 901 67
XM_017025910.1 XP_016881399.1 8 - 9 1480 67
XM_017025911.1 XP_016881400.1 8 - 9 1936 67
XM_017025912.1 XP_016881401.1 8 - 9 1480 67
GenBank mRNA AK294492.1 - 4 - 5 822 67
AK303652.1 - 8 - 9 1439 67
BC051651.1 - 8 - 9 1677 67
BC151842.1 - 8 - 9 2487 67
X58288.1 - 8 - 9 1438 67

View Full Product Details