Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
Chromosome Location
Chr.5: 13690328 - 14011829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001369.2 NP_001360.1 41 - 42 6881 65
XM_005248262.3 XP_005248319.2 41 - 42 6964 65
XM_017009177.1 XP_016864666.1 41 - 42 6964 65
XM_017009178.1 XP_016864667.1 41 - 42 7767 65
XM_017009179.1 XP_016864668.1 35 - 36 6061 65
XM_017009180.1 XP_016864669.1 41 - 42 6964 65
XM_017009181.1 XP_016864670.1 41 - 42 6964 65
XM_017009182.1 XP_016864671.1 41 - 42 6964 65
XM_017009183.1 XP_016864672.1 41 - 42 6964 65
XM_017009184.1 XP_016864673.1 41 - 42 6964 65
XM_017009185.1 XP_016864674.1 13 - 14 2260 65
XM_017009186.1 XP_016864675.1 10 - 11 1676 65
XM_017009187.1 XP_016864676.1 41 - 42 6964 65
XM_017009188.1 XP_016864677.1 7 - 8 2492 65
GenBank mRNA AY045575.1 - 41 - 42 6881 65
AY049075.1 - 6 - 7 974 65

View Full Product Details