Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH5
Entrez Gene ID
1767
Gene Name
dynein axonemal heavy chain 5
Gene Aliases
CILD3, DNAHC5, HL1, KTGNR, PCD
Chromosome Location
Chr.5: 13690328 - 14011829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001369.2 NP_001360.1 55 - 56 9410 86
XM_005248262.3 XP_005248319.2 55 - 56 9493 86
XM_017009177.1 XP_016864666.1 55 - 56 9493 86
XM_017009178.1 XP_016864667.1 55 - 56 10296 86
XM_017009179.1 XP_016864668.1 49 - 50 8590 86
XM_017009180.1 XP_016864669.1 55 - 56 9493 86
XM_017009181.1 XP_016864670.1 55 - 56 9493 86
XM_017009182.1 XP_016864671.1 55 - 56 9493 86
XM_017009183.1 XP_016864672.1 55 - 56 9493 86
XM_017009185.1 XP_016864674.1 27 - 28 4789 86
XM_017009186.1 XP_016864675.1 24 - 25 4205 86
XM_017009188.1 XP_016864677.1 21 - 22 5021 86
GenBank mRNA AB046823.2 - 3 - 4 474 86
AB290164.1 - 8 - 9 1413 86
AY045575.1 - 55 - 56 9410 86

View Full Product Details