The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282788.1 | NP_001269717.1 | 1 - 2 | 346 | 81 |
NM_032599.3 | NP_115988.1 | 1 - 2 | 346 | 81 | |
NR_104242.1 | 1 - 2 | 346 | 81 | ||
XM_017012743.1 | XP_016868232.1 | 1 - 2 | 389 | 81 | |
GenBank mRNA | AF367470.1 | - | 1 - 2 | 299 | 81 |
AK310308.1 | - | 1 - 2 | 293 | 81 | |
AK313608.1 | - | 1 - 2 | 297 | 81 | |
BC037248.1 | - | 1 - 2 | 285 | 81 | |
HY051242.1 | - | 1 - 2 | 349 | 81 |