The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206897.1 | NP_001193826.1 | 13 - 14 | 1858 | 97 |
NM_003888.3 | NP_003879.2 | 12 - 13 | 1761 | 97 | |
NM_170696.2 | NP_733797.1 | 11 - 12 | 1647 | 97 | |
NM_170697.2 | NP_733798.1 | 10 - 11 | 1365 | 97 | |
GenBank mRNA | AB015226.1 | - | 12 - 13 | 1531 | 97 |
AB015227.1 | - | 10 - 11 | 1294 | 97 | |
AB015228.1 | - | 10 - 11 | 1256 | 97 | |
AK128709.1 | - | 12 - 13 | 2256 | 97 | |
AK294981.1 | - | 12 - 13 | 1461 | 97 | |
AK303057.1 | - | 13 - 14 | 1603 | 97 | |
AL110299.1 | - | 4 - 5 | 1309 | 97 | |
BC030589.2 | - | 11 - 12 | 1405 | 97 |