The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025078.4 | NP_079354.2 | 4 - 5 | 570 | 54 |
XM_017025997.1 | XP_016881486.1 | 5 - 6 | 657 | 54 | |
XM_017025999.1 | XP_016881488.1 | 5 - 6 | 657 | 54 | |
XM_017026001.1 | XP_016881490.1 | 3 - 4 | 587 | 54 | |
GenBank mRNA | AK123870.1 | - | 4 - 5 | 441 | 54 |
BC030140.2 | - | 4 - 5 | 534 | 54 |