Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
Chromosome Location
Chr.18: 31376777 - 31415791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177986.4 NP_817123.1 13 - 14 2210 79
XM_011525827.1 XP_011524129.1 3 - 4 2125 79
GenBank mRNA AY177664.1 - 13 - 14 2210 79
AY227350.1 - 13 - 14 2075 79
BC132907.1 - 13 - 14 2098 79
BC132909.1 - 13 - 14 2098 79

View Full Product Details