The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001129758.1 | NP_001123230.1 | 3 - 4 | 713 | 66 |
NM_022357.3 | NP_071752.3 | 3 - 4 | 713 | 66 | |
GenBank mRNA | AJ291679.1 | - | 3 - 4 | 627 | 66 |
AK057401.1 | - | 3 - 4 | 713 | 66 | |
AY358390.1 | - | 3 - 4 | 636 | 66 | |
BC037243.1 | - | 3 - 4 | 625 | 66 | |
BC057789.1 | - | 3 - 4 | 654 | 66 |