The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002150.2 | NP_002141.1 | 1 - 2 | 37 | 69 |
GenBank mRNA | AK290826.1 | - | 1 - 2 | 33 | 69 |
CD675915.1 | - | 1 - 2 | 166 | 69 |