Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FTSJ1
Entrez Gene ID
24140
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
Chromosome Location
Chr.X: 48476021 - 48486364 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012280.3 NP_036412.1 7 - 8 927 102
NM_177439.2 NP_803188.1 8 - 9 1033 102
XM_005272595.1 XP_005272652.1 7 - 8 882 102
XM_011543893.2 XP_011542195.1 7 - 8 882 102
XM_011543894.2 XP_011542196.1 8 - 9 878 102
XM_011543895.1 XP_011542197.1 7 - 8 882 102
XM_017029382.1 XP_016884871.1 8 - 9 878 102
GenBank mRNA AF063015.1 - 6 - 7 520 102
AJ005892.1 - 7 - 8 764 102
AK024023.1 - 8 - 9 893 102
AK315138.1 - 7 - 8 709 102
BC021047.1 - 7 - 8 695 102
BC023584.1 - 7 - 8 693 102
KU178580.1 - 6 - 7 464 102
KU178581.1 - 6 - 7 464 102

View Full Product Details