The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001917.4 | NP_001908.3 | 9 - 10 | 963 | 65 |
XM_005268692.3 | XP_005268749.1 | 8 - 9 | 897 | 65 | |
XM_011538004.2 | XP_011536306.1 | 10 - 11 | 1049 | 65 | |
XM_011538005.2 | XP_011536307.1 | 10 - 11 | 1053 | 65 | |
GenBank mRNA | AK312995.1 | - | 9 - 10 | 950 | 65 |
AX741655.1 | - | 9 - 10 | 953 | 65 | |
AX741656.1 | - | 9 - 10 | 953 | 65 | |
AX741658.1 | - | 8 - 9 | 838 | 65 | |
AX741659.1 | - | 6 - 7 | 725 | 65 | |
AX776478.1 | - | 9 - 10 | 953 | 65 | |
AX776479.1 | - | 9 - 10 | 953 | 65 | |
AX776480.1 | - | 11 - 12 | 1882 | 65 | |
AX786386.1 | - | 9 - 10 | 953 | 65 | |
BC029057.1 | - | 9 - 10 | 933 | 65 | |
BC074770.2 | - | 9 - 10 | 835 | 65 | |
BX538210.1 | - | 9 - 10 | 895 | 65 | |
KU178015.1 | - | 8 - 9 | 810 | 65 |