The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018191.3 | NP_060661.3 | 11 - 12 | 1584 | 141 |
XM_005266441.2 | XP_005266498.1 | 10 - 11 | 1504 | 141 | |
XM_011535133.1 | XP_011533435.1 | 11 - 12 | 1610 | 141 | |
XM_011535134.1 | XP_011533436.1 | 11 - 12 | 1510 | 141 | |
XM_011535135.2 | XP_011533437.1 | 11 - 12 | 1468 | 141 | |
GenBank mRNA | AB209222.1 | - | 2 - 3 | 1713 | 141 |
AF334406.1 | - | 11 - 12 | 1558 | 141 | |
AJ319660.1 | - | 11 - 12 | 1525 | 141 | |
AK001578.1 | - | 3 - 4 | 460 | 141 | |
AK090807.1 | - | 5 - 6 | 704 | 141 | |
AK096654.1 | - | 11 - 12 | 1584 | 141 | |
AL833821.1 | - | 11 - 12 | 1494 | 141 |