The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018191.3 | NP_060661.3 | 8 - 9 | 1113 | 58 |
XM_005266441.2 | XP_005266498.1 | 7 - 8 | 1033 | 58 | |
XM_011535133.1 | XP_011533435.1 | 8 - 9 | 1139 | 58 | |
XM_011535134.1 | XP_011533436.1 | 8 - 9 | 1039 | 58 | |
XM_011535135.2 | XP_011533437.1 | 8 - 9 | 997 | 58 | |
GenBank mRNA | AF334406.1 | - | 8 - 9 | 1087 | 58 |
AJ319660.1 | - | 8 - 9 | 1054 | 58 | |
AK090807.1 | - | 2 - 3 | 233 | 58 | |
AK096654.1 | - | 8 - 9 | 1113 | 58 | |
AL833821.1 | - | 8 - 9 | 1023 | 58 | |
BC038104.2 | - | 8 - 9 | 1107 | 58 |