Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTIF3
Entrez Gene ID
219402
Gene Name
mitochondrial translational initiation factor 3
Gene Aliases
IF3mt
Chromosome Location
Chr.13: 27435639 - 27450900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006719771.3 XP_006719834.1 1 - 2 77 138

View Full Product Details