The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114331.2 | NP_001107803.1 | 10 - 11 | 1018 | 63 |
NM_001287.5 | NP_001278.1 | 11 - 12 | 1090 | 63 | |
XM_011522354.1 | XP_011520656.1 | 11 - 12 | 1172 | 63 | |
GenBank mRNA | AF224741.1 | - | 11 - 12 | 1018 | 63 |
AK056551.1 | - | 10 - 11 | 959 | 63 | |
AK291404.1 | - | 11 - 12 | 1033 | 63 | |
AK304796.1 | - | 11 - 12 | 883 | 63 | |
AK315978.1 | - | 12 - 13 | 913 | 63 | |
BC012737.2 | - | 11 - 12 | 1017 | 63 | |
Z67743.1 | - | 11 - 12 | 932 | 63 |