The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287.5 | NP_001278.1 | 2 - 3 | 329 | 77 |
XM_011522354.1 | XP_011520656.1 | 2 - 3 | 411 | 77 | |
GenBank mRNA | AF224741.1 | - | 2 - 3 | 257 | 77 |
AK122944.1 | - | 2 - 3 | 845 | 77 | |
AK291404.1 | - | 2 - 3 | 272 | 77 | |
AK304796.1 | - | 2 - 3 | 122 | 77 | |
AK315978.1 | - | 3 - 4 | 152 | 77 | |
BC012737.2 | - | 2 - 3 | 256 | 77 | |
DB061109.1 | - | 2 - 3 | 330 | 77 | |
Z67743.1 | - | 2 - 3 | 171 | 77 |