Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM57A
Entrez Gene ID
79850
Gene Name
family with sequence similarity 57 member A
Gene Aliases
CT120
Chromosome Location
Chr.17: 732546 - 742968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318006.1 NP_001304935.1 3 - 4 510 80
GenBank mRNA BC026023.1 - 3 - 4 472 80

View Full Product Details