The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016426.6 | NP_057510.4 | 10 - 11 | 2134 | 85 |
XM_005261627.3 | XP_005261684.1 | 10 - 11 | 2994 | 85 | |
XM_011530211.2 | XP_011528513.1 | 9 - 10 | 2936 | 85 | |
XM_017028815.1 | XP_016884304.1 | 9 - 10 | 2384 | 85 | |
GenBank mRNA | AF223408.1 | - | 9 - 10 | 1935 | 85 |
AK025563.1 | - | 10 - 11 | 1961 | 85 | |
AK222806.1 | - | 10 - 11 | 1961 | 85 | |
AK303087.1 | - | 9 - 10 | 1905 | 85 | |
AK315550.1 | - | 10 - 11 | 1960 | 85 | |
BC006325.2 | - | 9 - 10 | 1912 | 85 | |
BC075828.1 | - | 10 - 11 | 2035 | 85 | |
CT841519.1 | - | 9 - 10 | 1878 | 85 |