The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271213.1 | NP_001258142.1 | 7 - 8 | 1059 | 82 |
NM_021199.3 | NP_067022.1 | 6 - 7 | 1252 | 82 | |
GenBank mRNA | AF042284.1 | - | 6 - 7 | 942 | 82 |
AF118085.1 | - | 5 - 6 | 773 | 82 | |
AF151802.1 | - | 6 - 7 | 936 | 82 | |
AK130140.1 | - | 7 - 8 | 1059 | 82 | |
AK222463.1 | - | 6 - 7 | 951 | 82 | |
AK313111.1 | - | 6 - 7 | 931 | 82 | |
BC016836.1 | - | 6 - 7 | 907 | 82 |