Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YKT6
Entrez Gene ID
10652
Gene Name
YKT6 v-SNARE homolog (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.7: 44200968 - 44214294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006555.3 NP_006546.1 5 - 6 611 74
XM_005249582.4 XP_005249639.1 5 - 6 612 74
GenBank mRNA AB451317.1 - 5 - 6 454 74
AB451447.1 - 5 - 6 454 74
AF091079.1 - 5 - 6 514 74
AK223488.1 - 5 - 6 568 74
AK312622.1 - 5 - 6 600 74
BC007319.2 - 5 - 6 541 74
BT007078.1 - 5 - 6 454 74
CR456989.1 - 5 - 6 454 74
CR542008.1 - 5 - 6 454 74
U95735.1 - 5 - 6 513 74

View Full Product Details