The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199771.1 | NP_001186700.1 | 1 - 2 | 146 | 71 |
GenBank mRNA | BC028298.1 | - | 1 - 2 | 100 | 71 |
BI765016.1 | - | 1 - 2 | 52 | 71 |