The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178078.1 | NP_001171549.1 | 4 - 5 | 670 | 120 |
NM_001178079.1 | NP_001171550.1 | 4 - 5 | 596 | 120 | |
NM_001178080.1 | NP_001171551.1 | 3 - 4 | 514 | 120 | |
NM_001178081.1 | NP_001171552.1 | 2 - 3 | 375 | 120 | |
NM_003153.4 | NP_003144.3 | 4 - 5 | 651 | 120 | |
NR_033659.1 | 3 - 4 | 512 | 120 | ||
XM_006719575.2 | XP_006719638.1 | 5 - 6 | 648 | 120 | |
XM_011538703.2 | XP_011537005.1 | 4 - 5 | 671 | 120 | |
XM_011538704.2 | XP_011537006.1 | 5 - 6 | 682 | 120 | |
XM_011538705.2 | XP_011537007.1 | 5 - 6 | 792 | 120 | |
XM_011538706.2 | XP_011537008.1 | 5 - 6 | 648 | 120 | |
XM_011538707.2 | XP_011537009.1 | 4 - 5 | 633 | 120 | |
XM_011538708.2 | XP_011537010.1 | 3 - 4 | 475 | 120 | |
XM_011538709.2 | XP_011537011.1 | 4 - 5 | 509 | 120 | |
GenBank mRNA | AK290431.1 | - | 4 - 5 | 599 | 120 |
AK291044.1 | - | 4 - 5 | 597 | 120 | |
AK298721.1 | - | 2 - 3 | 195 | 120 | |
AK299094.1 | - | 3 - 4 | 448 | 120 | |
AK308751.1 | - | 4 - 5 | 494 | 120 | |
AK312471.1 | - | 4 - 5 | 517 | 120 | |
AK316142.1 | - | 3 - 4 | 458 | 120 | |
AK316236.1 | - | 3 - 4 | 289 | 120 | |
BC075852.1 | - | 4 - 5 | 452 | 120 | |
BT007380.1 | - | 3 - 4 | 345 | 120 | |
U16031.1 | - | 4 - 5 | 510 | 120 |