Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STAT6
Entrez Gene ID
6778
Gene Name
signal transducer and activator of transcription 6
Gene Aliases
D12S1644, IL-4-STAT, STAT6B, STAT6C
Chromosome Location
Chr.12: 57095404 - 57111433 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178078.1 NP_001171549.1 8 - 9 1137 92
NM_001178079.1 NP_001171550.1 8 - 9 1063 92
NM_001178080.1 NP_001171551.1 7 - 8 981 92
NM_001178081.1 NP_001171552.1 6 - 7 842 92
NM_003153.4 NP_003144.3 8 - 9 1118 92
NR_033659.1 7 - 8 979 92
XM_006719575.2 XP_006719638.1 9 - 10 1115 92
XM_011538703.2 XP_011537005.1 8 - 9 1138 92
XM_011538704.2 XP_011537006.1 9 - 10 1149 92
XM_011538705.2 XP_011537007.1 9 - 10 1259 92
XM_011538706.2 XP_011537008.1 9 - 10 1115 92
XM_011538707.2 XP_011537009.1 8 - 9 1100 92
XM_011538708.2 XP_011537010.1 7 - 8 942 92
XM_011538709.2 XP_011537011.1 8 - 9 976 92
GenBank mRNA AB103089.1 - 2 - 3 290 92
AK290431.1 - 8 - 9 1066 92
AK291044.1 - 8 - 9 1064 92
AK298721.1 - 6 - 7 662 92
AK299094.1 - 7 - 8 915 92
AK308751.1 - 8 - 9 961 92
AK316142.1 - 7 - 8 925 92
AK316236.1 - 7 - 8 756 92
BC075852.1 - 8 - 9 919 92
BT007380.1 - 7 - 8 812 92
U16031.1 - 8 - 9 977 92

View Full Product Details