The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020470.2 | NP_065203.2 | 5 - 6 | 667 | 102 |
XM_005273720.3 | XP_005273777.1 | 5 - 6 | 643 | 102 | |
GenBank mRNA | AK226003.1 | - | 5 - 6 | 645 | 102 |
AK291124.1 | - | 5 - 6 | 645 | 102 | |
BC001299.1 | - | 5 - 6 | 618 | 102 | |
BC009892.2 | - | 2 - 3 | 94 | 102 | |
CR456710.1 | - | 5 - 6 | 485 | 102 |