The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014247.2 | NP_055062.1 | 14 - 15 | 2746 | 90 |
XM_005263358.3 | XP_005263415.1 | 20 - 21 | 3013 | 90 | |
XM_005263359.3 | XP_005263416.1 | 20 - 21 | 3015 | 90 | |
XM_005263360.3 | XP_005263417.1 | 20 - 21 | 3007 | 90 | |
XM_005263361.3 | XP_005263418.1 | 19 - 20 | 2995 | 90 | |
XM_006714420.3 | XP_006714483.1 | 19 - 20 | 2995 | 90 | |
XM_006714421.3 | XP_006714484.1 | 20 - 21 | 2980 | 90 | |
XM_006714422.3 | XP_006714485.1 | 17 - 18 | 3228 | 90 | |
XM_011532425.2 | XP_011530727.1 | 16 - 17 | 2843 | 90 | |
XM_011532426.2 | XP_011530728.1 | 17 - 18 | 3780 | 90 | |
XM_011532427.2 | XP_011530729.1 | 14 - 15 | 3430 | 90 | |
XM_017008856.1 | XP_016864345.1 | 20 - 21 | 3014 | 90 | |
XM_017008857.1 | XP_016864346.1 | 17 - 18 | 3779 | 90 | |
XM_017008858.1 | XP_016864347.1 | 17 - 18 | 2747 | 90 | |
XM_017008859.1 | XP_016864348.1 | 14 - 15 | 3430 | 90 | |
GenBank mRNA | AB002311.1 | - | 14 - 15 | 2498 | 90 |
AK294459.1 | - | 14 - 15 | 2747 | 90 | |
BC117321.1 | - | 14 - 15 | 2424 | 90 |