Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAPGEF2
Entrez Gene ID
9693
Gene Name
Rap guanine nucleotide exchange factor 2
Gene Aliases
CNrasGEF, NRAPGEP, PDZ-GEF1, PDZGEF1, RA-GEF, RA-GEF-1, Rap-GEP, nRap GEP
Chromosome Location
Chr.4: 159104067 - 159360164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014247.2 NP_055062.1 5 - 6 972 116
XM_005263358.3 XP_005263415.1 11 - 12 1239 116
XM_005263359.3 XP_005263416.1 11 - 12 1241 116
XM_005263360.3 XP_005263417.1 11 - 12 1233 116
XM_005263361.3 XP_005263418.1 10 - 11 1221 116
XM_006714420.3 XP_006714483.1 10 - 11 1221 116
XM_006714421.3 XP_006714484.1 11 - 12 1206 116
XM_006714422.3 XP_006714485.1 8 - 9 1454 116
XM_011532425.2 XP_011530727.1 7 - 8 1069 116
XM_011532426.2 XP_011530728.1 8 - 9 2006 116
XM_011532427.2 XP_011530729.1 5 - 6 1656 116
XM_017008856.1 XP_016864345.1 11 - 12 1240 116
XM_017008857.1 XP_016864346.1 8 - 9 2005 116
XM_017008858.1 XP_016864347.1 8 - 9 973 116
XM_017008859.1 XP_016864348.1 5 - 6 1656 116
GenBank mRNA AB002311.1 - 5 - 6 724 116
AK294459.1 - 5 - 6 973 116
BC117321.1 - 5 - 6 650 116

View Full Product Details