The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144823.2 | NP_001138295.1 | 12 - 13 | 1974 | 121 |
NM_001320835.1 | NP_001307764.1 | 12 - 13 | 1974 | 121 | |
NM_005848.4 | NP_005839.3 | 12 - 13 | 1967 | 121 | |
XM_005254120.4 | XP_005254177.1 | 11 - 12 | 1753 | 121 | |
XM_005254121.3 | XP_005254178.1 | 12 - 13 | 1974 | 121 | |
XM_005254122.4 | XP_005254179.1 | 12 - 13 | 1833 | 121 | |
XM_006720371.3 | XP_006720434.1 | 12 - 13 | 1709 | 121 | |
XM_011521156.2 | XP_011519458.1 | 12 - 13 | 1969 | 121 | |
XM_011521157.2 | XP_011519459.1 | 2 - 3 | 309 | 121 | |
XM_017021862.1 | XP_016877351.1 | 11 - 12 | 1694 | 121 | |
XM_017021863.1 | XP_016877352.1 | 11 - 12 | 1752 | 121 | |
XM_017021864.1 | XP_016877353.1 | 12 - 13 | 1969 | 121 | |
XM_017021865.1 | XP_016877354.1 | 12 - 13 | 1832 | 121 | |
GenBank mRNA | AF534403.1 | - | 12 - 13 | 1929 | 121 |
AK091368.1 | - | 12 - 13 | 1929 | 121 | |
AK290292.1 | - | 12 - 13 | 1782 | 121 | |
AK299570.1 | - | 12 - 13 | 1888 | 121 | |
AK307411.1 | - | 12 - 13 | 1974 | 121 | |
AL832602.2 | - | 10 - 11 | 1493 | 121 | |
AL833317.1 | - | 12 - 13 | 1805 | 121 | |
AL833384.1 | - | 9 - 10 | 1238 | 121 | |
BC028057.1 | - | 12 - 13 | 1819 | 121 | |
BC041706.1 | - | 12 - 13 | 1798 | 121 |