The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144823.2 | NP_001138295.1 | 8 - 9 | 1494 | 133 |
NM_001320835.1 | NP_001307764.1 | 8 - 9 | 1494 | 133 | |
NM_005848.4 | NP_005839.3 | 8 - 9 | 1487 | 133 | |
XM_005254120.4 | XP_005254177.1 | 7 - 8 | 1273 | 133 | |
XM_005254121.3 | XP_005254178.1 | 8 - 9 | 1494 | 133 | |
XM_005254122.4 | XP_005254179.1 | 8 - 9 | 1353 | 133 | |
XM_006720371.3 | XP_006720434.1 | 8 - 9 | 1229 | 133 | |
XM_011521156.2 | XP_011519458.1 | 8 - 9 | 1489 | 133 | |
XM_017021862.1 | XP_016877351.1 | 7 - 8 | 1214 | 133 | |
XM_017021863.1 | XP_016877352.1 | 7 - 8 | 1272 | 133 | |
XM_017021864.1 | XP_016877353.1 | 8 - 9 | 1489 | 133 | |
XM_017021865.1 | XP_016877354.1 | 8 - 9 | 1352 | 133 | |
GenBank mRNA | AF534403.1 | - | 8 - 9 | 1449 | 133 |
AK091368.1 | - | 8 - 9 | 1449 | 133 | |
AK290292.1 | - | 8 - 9 | 1302 | 133 | |
AK299570.1 | - | 8 - 9 | 1408 | 133 | |
AK307411.1 | - | 8 - 9 | 1494 | 133 | |
AL833384.1 | - | 5 - 6 | 758 | 133 | |
BC028057.1 | - | 8 - 9 | 1339 | 133 | |
BC041706.1 | - | 8 - 9 | 1318 | 133 |