Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM5
Entrez Gene ID
10329
Gene Name
transmembrane protein 5
Gene Aliases
HP10481, MDDGA10
Chromosome Location
Chr.12: 63779803 - 63809558 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278237.1 NP_001265166.1 3 - 4 595 89
NM_014254.2 NP_055069.1 3 - 4 585 89
XM_005268562.3 XP_005268619.1 4 - 5 695 89
XM_005268563.3 XP_005268620.1 4 - 5 768 89
GenBank mRNA AB015633.1 - 3 - 4 531 89
AK289382.1 - 3 - 4 479 89
BC002596.2 - 3 - 4 516 89
BC013152.1 - 3 - 4 517 89
BM547839.1 - 3 - 4 595 89
BX647105.1 - 3 - 4 584 89
BX647419.1 - 3 - 4 584 89

View Full Product Details