The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004946.2 | NP_004937.1 | 31 - 32 | 3259 | 77 |
GenBank mRNA | AK128107.1 | - | 17 - 18 | 1820 | 77 |
BC104900.1 | - | 31 - 32 | 3215 | 77 | |
BC113457.1 | - | 31 - 32 | 3215 | 77 | |
BC143358.1 | - | 32 - 33 | 3266 | 77 | |
D86964.1 | - | 31 - 32 | 3217 | 77 |