The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004946.2 | NP_004937.1 | 40 - 41 | 4151 | 63 |
GenBank mRNA | AK128107.1 | - | 26 - 27 | 2712 | 63 |
BC084556.1 | - | 5 - 6 | 432 | 63 | |
BC104900.1 | - | 40 - 41 | 4107 | 63 | |
BC113457.1 | - | 40 - 41 | 4107 | 63 | |
BC143358.1 | - | 41 - 42 | 4158 | 63 | |
D86964.1 | - | 40 - 41 | 4109 | 63 |