The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014692.1 | NP_055507.1 | 12 - 13 | 1610 | 122 |
GenBank mRNA | AB007880.2 | - | 12 - 13 | 1610 | 122 |