The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139076.2 | NP_620775.2 | 1 - 2 | 155 | 68 |
XM_017008694.1 | XP_016864183.1 | 1 - 2 | 128 | 68 | |
GenBank mRNA | AK022704.1 | - | 1 - 2 | 141 | 68 |
AK023676.1 | - | 1 - 2 | 122 | 68 | |
AY358576.1 | - | 1 - 2 | 106 | 68 | |
BC016905.1 | - | 1 - 2 | 83 | 68 | |
BC039573.1 | - | 1 - 2 | 102 | 68 | |
DA765424.1 | - | 1 - 2 | 155 | 68 | |
EF531340.1 | - | 1 - 2 | 90 | 68 |