Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSPO
Entrez Gene ID
23145
Gene Name
SCO-spondin
Gene Aliases
-
Chromosome Location
Chr.7: 149776042 - 149833965 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198455.2 NP_940857.2 67 - 68 10176 134

View Full Product Details