Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TATDN3
Entrez Gene ID
128387
Gene Name
TatD DNase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 212791828 - 212816825 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042552.2 NP_001036017.1 2 - 3 194 68
NM_001042553.2 NP_001036018.1 2 - 3 194 68
NM_001146169.1 NP_001139641.1 2 - 3 194 68
NM_001146170.1 NP_001139642.1 2 - 3 194 68
NM_001146171.1 NP_001139643.1 2 - 3 194 68
XM_011509202.2 XP_011507504.1 2 - 3 189 68
XM_017000328.1 XP_016855817.1 2 - 3 189 68
XM_017000329.1 XP_016855818.1 2 - 3 189 68
XM_017000330.1 XP_016855819.1 2 - 3 189 68
XM_017000331.1 XP_016855820.1 2 - 3 189 68
GenBank mRNA AK293129.1 - 2 - 3 194 68
AK304557.1 - 2 - 3 152 68
AL832248.1 - 2 - 3 136 68
BC048115.1 - 2 - 3 105 68
BC113638.1 - 2 - 3 138 68
BC117485.1 - 2 - 3 138 68
BC143956.1 - 2 - 3 138 68
BC143957.1 - 2 - 3 138 68
DB456872.1 - 2 - 3 172 68

View Full Product Details