Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TATDN3
Entrez Gene ID
128387
Gene Name
TatD DNase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.1: 212791828 - 212816825 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146169.1 NP_001139641.1 8 - 9 693 156
NM_001146171.1 NP_001139643.1 8 - 9 693 156
XM_017000328.1 XP_016855817.1 7 - 8 625 156
GenBank mRNA AK293129.1 - 8 - 9 693 156
BC143957.1 - 8 - 9 637 156

View Full Product Details