Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHCHD10
Entrez Gene ID
400916
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Gene Aliases
C22orf16, FTDALS2, IMMD, N27C7-4, SMAJ
Chromosome Location
Chr.22: 23765834 - 23767972 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213720.2 NP_998885.1 2 - 3 361 131
NR_125755.1 2 - 3 308 131
GenBank mRNA AK289560.1 - 2 - 3 340 131
BC053570.1 - 2 - 3 312 131
BC065232.1 - 2 - 3 343 131
CN273854.1 - 2 - 3 256 131

View Full Product Details