The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039948.3 | NP_001035037.1 | 5 - 6 | 556 | 79 |
NM_001098497.2 | NP_001091967.1 | 5 - 6 | 556 | 79 | |
NM_001098498.2 | NP_001091968.1 | 5 - 6 | 556 | 79 | |
NM_133454.3 | NP_597711.1 | 5 - 6 | 556 | 79 | |
GenBank mRNA | AB075821.1 | - | - | 1825 | 79 |
AB275761.1 | - | 5 - 6 | 556 | 79 | |
AB449879.1 | - | - | 376 | 79 |