The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085447.1 | NP_001078916.1 | 2 - 3 | 388 | 99 |
XM_011510589.1 | XP_011508891.1 | 2 - 3 | 276 | 99 | |
XM_011510590.1 | XP_011508892.1 | 2 - 3 | 309 | 99 | |
GenBank mRNA | AK301851.1 | - | 2 - 3 | 356 | 99 |
BC117445.1 | - | 2 - 3 | 331 | 99 | |
BQ432665.1 | - | 2 - 3 | 390 | 99 |