Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC173
Entrez Gene ID
129881
Gene Name
coiled-coil domain containing 173
Gene Aliases
C2orf77
Chromosome Location
Chr.2: 169645425 - 169694421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085447.1 NP_001078916.1 4 - 5 760 105
XM_011510589.1 XP_011508891.1 4 - 5 648 105
XM_011510590.1 XP_011508892.1 4 - 5 681 105
GenBank mRNA AK301851.1 - 4 - 5 728 105
BC117445.1 - 4 - 5 703 105

View Full Product Details