Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LINC00475
Entrez Gene ID
158314
Gene Name
long intergenic non-protein coding RNA 475
Gene Aliases
C9orf44
Chromosome Location
Chr.9: 92141467 - 92159608 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027341.1 1 - 2 179 77
GenBank mRNA AK023662.1 - 1 - 2 179 77

View Full Product Details