Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf57
Entrez Gene ID
138240
Gene Name
chromosome 9 open reading frame 57
Gene Aliases
-
Chromosome Location
Chr.9: 72051381 - 72060605 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128618.1 NP_001122090.1 1 - 2 113 88
GenBank mRNA BC036255.1 - 1 - 2 113 88
BC044620.1 - 1 - 2 91 88
BC053956.1 - 1 - 2 91 88

View Full Product Details