The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207363.2 | NP_997246.2 | 7 - 8 | 804 | 97 |
NM_207481.3 | NP_997364.3 | 7 - 8 | 804 | 97 | |
XM_005263659.3 | XP_005263716.1 | 6 - 7 | 1386 | 97 | |
XM_005263660.3 | XP_005263717.1 | 6 - 7 | 901 | 97 | |
XM_011511097.2 | XP_011509399.1 | 8 - 9 | 1574 | 97 | |
XM_011511098.2 | XP_011509400.1 | 8 - 9 | 1571 | 97 | |
XM_011511099.2 | XP_011509401.1 | 7 - 8 | 969 | 97 | |
XM_011511100.2 | XP_011509402.1 | 7 - 8 | 1067 | 97 | |
XM_011511101.1 | XP_011509403.1 | 5 - 6 | 477 | 97 | |
XM_011511102.2 | XP_011509404.1 | 6 - 7 | 4903 | 97 | |
XM_011511103.2 | XP_011509405.1 | 4 - 5 | 705 | 97 | |
XM_011511104.1 | XP_011509406.1 | 4 - 5 | 356 | 97 | |
XM_011511105.2 | XP_011509407.1 | 4 - 5 | 1147 | 97 | |
XM_017003974.1 | XP_016859463.1 | 7 - 8 | 697 | 97 | |
XM_017003975.1 | XP_016859464.1 | 6 - 7 | 999 | 97 | |
XM_017003976.1 | XP_016859465.1 | 8 - 9 | 1595 | 97 | |
XM_017003977.1 | XP_016859466.1 | 5 - 6 | 1505 | 97 | |
XM_017003979.1 | XP_016859468.1 | 7 - 8 | 1444 | 97 | |
XM_017003980.1 | XP_016859469.1 | 4 - 5 | 705 | 97 | |
GenBank mRNA | AK092189.1 | - | 5 - 6 | 479 | 97 |
AY946007.1 | - | 2 - 3 | 226 | 97 | |
AY946008.1 | - | 2 - 3 | 226 | 97 |