Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
Chromosome Location
Chr.1: 68474152 - 68497221 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114120.2 NP_001107592.1 11 - 12 2515 150
NM_017779.5 NP_060249.2 10 - 11 1663 150
GenBank mRNA AB281187.1 - 11 - 12 2371 150
AB281274.1 - 10 - 11 1519 150
AJ278112.1 - 10 - 11 1519 150
AK000490.1 - 4 - 5 840 150
AK022792.1 - 3 - 4 1156 150
AK225735.1 - 11 - 12 2361 150
AK299324.1 - 11 - 12 2376 150
AL834208.1 - 12 - 13 1435 150
BC065304.1 - 10 - 11 1466 150

View Full Product Details