The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114120.2 | NP_001107592.1 | 4 - 5 | 810 | 147 |
NM_017779.5 | NP_060249.2 | 4 - 5 | 810 | 147 | |
GenBank mRNA | AB281187.1 | - | 4 - 5 | 666 | 147 |
AB281274.1 | - | 4 - 5 | 666 | 147 | |
AJ278112.1 | - | 4 - 5 | 666 | 147 | |
AK000361.1 | - | 4 - 5 | 671 | 147 | |
AK225735.1 | - | 4 - 5 | 656 | 147 | |
AK299324.1 | - | 4 - 5 | 671 | 147 | |
AL834208.1 | - | 4 - 5 | 292 | 147 | |
BC003511.1 | - | 4 - 5 | 659 | 147 | |
BC065304.1 | - | 4 - 5 | 613 | 147 |