Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM2-AS
Entrez Gene ID
101928607
Gene Name
TRPM2 antisense RNA
Gene Aliases
TRPM2-AS1
Chromosome Location
Chr.21: 44414590 - 44425563 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_109964.1 2 - 3 519 82
GenBank mRNA EU362988.1 - 2 - 3 519 82

View Full Product Details