The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013642.2 | NP_001013664.1 | 1 - 2 | 911 | 97 |
GenBank mRNA | AI366714.1 | - | 1 - 2 | 305 | 97 |
BC069216.1 | - | 1 - 2 | 908 | 97 |