The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017017178.1 | XP_016872667.1 | 27 - 28 | 13317 | 93 |
XM_017017179.1 | XP_016872668.1 | 28 - 29 | 13539 | 93 | |
XM_017017181.1 | XP_016872670.1 | 26 - 27 | 13247 | 93 | |
XM_017017185.1 | XP_016872674.1 | 26 - 27 | 11143 | 93 | |
XM_017017186.1 | XP_016872675.1 | 26 - 27 | 10431 | 93 | |
XM_017017187.1 | XP_016872676.1 | 26 - 27 | 14789 | 93 | |
GenBank mRNA | AB076400.1 | - | 9 - 10 | 2640 | 93 |