The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142685.1 | NP_001136157.1 | 3 - 4 | 66 | |
GenBank mRNA | AL833062.2 | - | 2 - 3 | 66 | |
CK823137.1 | - | 2 - 3 | 359 | 66 | |
EF127492.1 | - | 3 - 4 | 66 |